Assay Details
Gene Symbol: Gm31819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141885.1 | - | 1 - 2 | 69 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | deleted in colorectal carcinoma | Dcc |
C030036D22Rik, Igdcc1 |
-365.8kb | Sense | Chr. 18 - 71253613 - 72351231 on NCBI Build GRCm38 |
Downstream |
predicted gene, 31908 |
Gm31908 | 183.8kb | Sense | Chr. 18 - 72908890 - 72921090 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
