Assay Details
Gene Symbol: Gm32335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141974.1 | - | 1 - 2 | 2702 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger and BTB domain containing 16 | Zbtb16 |
AI467657, PLZF, Zfp145, lu |
-60.1kb | Anti-sense | Chr. 9 - 48651797 - 48836381 on NCBI Build GRCm38 |
Downstream | 5-hydroxytryptamine (serotonin) receptor 3B | Htr3b | 5-HT3B | 29.0kb | Anti-sense | Chr. 9 - 48935008 - 48964990 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
