Assay Details
Gene Symbol: Gm31908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK145281.1 | - | 3 - 4 | 360 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31819 |
Gm31819 | -183.8kb | Sense | Chr. 18 - 72686426 - 72725083 on NCBI Build GRCm38 | |
Downstream | mex3 homolog C (C. elegans) | Mex3c |
A130001D14Rik, BC035207, BM-013, Rkhd2 |
659.4kb | Anti-sense | Chr. 18 - 73572585 - 73592578 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
