Assay Details
Gene Symbol: Gm30918 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155516.1 | - | 1 - 2 | 52 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 29590 | Gm29590 | -28.9kb | Sense | Chr. 13 - 37508243 - 37512421 on NCBI Build GRCm38 | |
Downstream | expressed sequence AI463229 | AI463229 | 9130430E04 | 113.8kb | Anti-sense | Chr. 13 - 37657013 - 37670077 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
