Assay Details
Gene Symbol: LOC101056071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138601.1 | - | 3 | 2049 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Abelson helper integration site 1 | Ahi1 |
1700015F03Rik, Ahi-1, D10Bwg0629e |
-72.2kb | Sense | Chr. 10 - 20952489 - 21080429 on NCBI Build GRCm38 |
Downstream |
predicted gene, 33619 |
Gm33619 | 32.5kb | Sense | Chr. 10 - 21186921 - 21188618 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
