Assay Details
Gene Symbol: Ccdc138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162956.1 | NP_001156428.1 | 10 - 11 | 1123 | 60 | |
GenBank mRNA | BC119028.1 | - | 10 - 11 | 1112 | 8733777 | 60 |
BC119030.1 | - | 10 - 11 | 1112 | 8733779 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
