Assay Details
Gene Symbol: Hoxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079869.1 | NP_001073338.1 | 3 - 4 | 1003 | 198 | |
NM_010458.2 | NP_034588.2 | 1 - 2 | 926 | 198 | ||
GenBank mRNA | S35628.1 | - | 1 - 2 | 926 | 198 | |
S35738.1 | - | 3 - 4 | 1004 | 198 | ||
X66177.1 | - | 1 - 2 | 926 | 198 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
