Assay Details
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008077.4 | NP_032103.2 | 10 - 11 | 1236 | 89 | |
GenBank mRNA | AF326547.1 | - | 10 - 11 | 1186 | 89 | |
AF326548.1 | - | 10 - 11 | 1186 | 89 | ||
AF483492.1 | - | 9 - 10 | 1003 | 89 | ||
AF483493.1 | - | 9 - 10 | 1003 | 89 | ||
AK136801.1 | - | 10 - 11 | 1221 | 89 | ||
AK148497.1 | - | 10 - 11 | 1236 | 89 | ||
AK158827.1 | - | 9 - 10 | 1002 | 89 | ||
AK160410.1 | - | 9 - 10 | 1145 | 88 | ||
AK160868.1 | - | 10 - 11 | 1234 | 89 | ||
BC027059.1 | - | 10 - 11 | 1187 | 5358787 | 89 | |
KC134210.1 | - | 9 - 10 | 1003 | 89 | ||
S67453.1 | - | 5 - 6 | 412 | 89 | ||
Y12257.1 | - | 9 - 10 | 1003 | 89 | ||
Z49976.1 | - | 10 - 11 | 1123 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
