Assay Details
Gene Symbol: Rasa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009025.2 | NP_033051.2 | 14 - 15 | 1541 | 85 | |
GenBank mRNA | AK053353.1 | - | 14 - 15 | 1537 | 85 | |
AK141511.1 | - | 14 - 15 | 1548 | 85 | ||
AK147896.1 | - | 14 - 15 | 1509 | 85 | ||
AK166354.1 | - | 14 - 15 | 1539 | 85 | ||
BC057300.1 | - | 14 - 15 | 1532 | 6821254 | 85 | |
BC060195.1 | - | 14 - 15 | 1556 | 85 | ||
BC068297.1 | - | 14 - 15 | 1405 | 30102545 | 85 | |
U20238.1 | - | 14 - 15 | 1526 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
