Assay Details
Gene Symbol: Cdc42ep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027219.3 | NP_081495.1 | 1 - 2 | 79 | 109 | |
GenBank mRNA | AK007896.1 | - | 1 - 2 | 80 | 109 | |
BC016250.1 | - | 1 - 2 | 67 | 4910994 | 109 | |
BC083130.1 | - | 1 - 2 | 72 | 6441478 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007896.1, BC016250.1, BC083130.1, NM_027219.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
