Assay Details
Gene Symbol: Dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139139.2 | NP_631878.2 | 10 - 11 | 824 | 97 | |
GenBank mRNA | AK077385.1 | - | 10 - 11 | 784 | 97 | |
BC013487.1 | - | 10 - 11 | 803 | 4211118 | 97 | |
BP770408.1 | - | 1 - 2 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568392 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568391 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568390 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568394 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568393 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960616 | BC013487.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81624 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239795 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368512 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790382 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21780 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404779 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943457 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222927 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420275 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471435 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271826 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723378 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542837 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877282 | AK084440.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380776 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132362 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890620 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763355 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18841 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113128 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94655 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158836 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148019 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960616 | BC013487.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568388 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568375 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568383 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568387 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568374 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568381 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568379 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568392 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568386 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158836 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00473793 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00473766 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148019 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04664536 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00473779 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04664485 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00473772 | - | - |
Whole Mouse Genome G4122A | A_52_P521419 | AK077385.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P521419 | BC013487.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P521419 | BP770408.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P521419 | NM_139139.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P197415 | AK006322.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113458 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120535 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148019 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00473766 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158836 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04664536 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01714771 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01714761 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01714766 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01714764 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568391 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568390 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568394 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568393 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568383 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01890168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02277772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01825736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01653646 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01357220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02278089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01993940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01753344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01559659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01653645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01939723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01824457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01993941 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01568201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01559663 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02091965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02278086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01890162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01251385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02277770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01559658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21568310 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10486187 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10486190 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10486189 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10486188 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10486186 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449756_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420165_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10486185 | - | - |
Mouse Genome 430 2.0 Array | 1420165_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1446928_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449756_at | - | - |
Mouse Genome 430 2.0 Array | 1446928_at | - | - |
Mouse Genome 430 2.0 Array | 1449756_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420165_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69408_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69408_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200036957.mm.1 | TC0200004424.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200036957.mm.1 | NM_139139.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200036957.mm.1 | NM_139139.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200036959.mm.1 | TC0200004424.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200036959.mm.1 | NM_139139.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200036959.mm.1 | NM_139139.2 | Refseq |
Mouse Expression Set 430 | 1446928_at | - | - |
Mouse Expression Set 430 | 1446928_PM_at | - | - |
Murine Genome U74v2 Set | 160730_at | - | - |
Murine Genome U74v2 Set | 160730_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420165_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449756_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449756_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420165_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 512379 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2350750 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2350750 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002350750 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708920 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2708920 | - | - |
mouse-6 v1.1 (Illumina) | scl069408.2_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2350750 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl069408.2_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350750 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2708920 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708920 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl069408.2_13-S | - | - |
