Assay Details
Gene Symbol: 9130023H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177001.3 | NP_795975.1 | 1 | 1551 | 103 | |
GenBank mRNA | AK033604.1 | - | 1 | 1511 | 103 | |
AK084086.1 | - | 1 | 1554 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80414 | AK033604.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80414 | AK084086.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80414 | NM_177001.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80414 | AK033604.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80414 | AK084086.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80414 | NM_177001.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282276 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813574 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80414 | AK033604.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80414 | AK084086.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80414 | NM_177001.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80414 | AK033604.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80414 | AK084086.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80414 | NM_177001.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80414 | AK033604.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80414 | AK084086.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80414 | NM_177001.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80414 | AK033604.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80414 | AK084086.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80414 | NM_177001.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P309170 | BB040432.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P309170 | AK033604.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P309170 | AK084086.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P309170 | NM_177001.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136059 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509728 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06560490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509608 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06525385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06599544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06188108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06525388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06274923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06274921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06560499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06599561 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06599549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06560484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25509620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06584006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10568375 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456331_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10568374 | - | - |
Mouse Genome 430 2.0 Array | 1456331_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456331_at | - | - |
Mouse Expression Set 430 | 1456331_at | - | - |
Mouse Expression Set 430 | 1456331_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456331_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456331_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 501116 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1850131 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001850131 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631215 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2631215 | - | - |
mouse-6 v1.1 (Illumina) | scl30619.1.4_211-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1850131 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30619.1.4_211-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850131 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631218 | - | - |
