Assay Details
Gene Symbol: Wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146298.2 | NP_001139770.1 | 4 - 5 | 761 | 72 | |
NM_001282093.1 | NP_001269022.1 | 4 - 5 | 602 | 72 | ||
NM_153085.4 | NP_694725.3 | 4 - 5 | 761 | 72 | ||
GenBank mRNA | AF320996.1 | - | 4 - 5 | 541 | 72 | |
AK087826.1 | - | 4 - 5 | 518 | 72 | ||
AK156780.1 | - | 4 - 5 | 586 | 72 | ||
AK172416.1 | - | 4 - 5 | 585 | 72 | ||
AK173274.1 | - | 4 - 5 | 463 | 72 | ||
BC020146.1 | - | 4 - 5 | 573 | 72 | ||
BC057178.1 | - | 4 - 5 | 394 | 72 | ||
BC080851.1 | - | 4 - 5 | 719 | 6848738 | 72 | |
BF461147.1 | - | 4 - 5 | 429 | 72 | ||
CA561963.1 | - | 2 - 3 | 216 | 72 | ||
CB249484.1 | - | 4 - 5 | 576 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
