Assay Details
Gene Symbol: Mir221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029806.1 | 1 | 43 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930578C19 gene | 4930578C19Rik | -684.9kb | Sense | Chr. X - 18414167 - 18461421 on NCBI Build GRCm38 | |
Downstream | microRNA 222 | Mir222 |
Mirn222, mir-222, mmu-mir-222 |
505.0 | Sense | Chr. X - 19146893 - 19146971 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
