Assay Details
Gene Symbol: 1700111N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033213.2 | 7 - 8 | 654 | 94 | ||
GenBank mRNA | AK007172.1 | - | 7 - 8 | 655 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700020N15 gene | 1700020N15Rik | -3.1kb | Anti-sense | Chr. X - 69945281 - 69945980 on NCBI Build GRCm38 | |
Downstream | iduronate 2-sulfatase | Ids | AW214631 | 375.5kb | Sense | Chr. X - 70343069 - 70365114 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
