Assay Details
Gene Symbol: 1700111N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033213.2 | 8 - 9 | 702 | 76 | ||
GenBank mRNA | AK007172.1 | - | 8 - 9 | 703 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
AF4/FMR2 family, member 2 |
Aff2 |
FMR2P, Fmr2, Ox19, Oxh |
-57.2kb | Anti-sense | Chr. X - 69360331 - 69872054 on NCBI Build GRCm38 |
Downstream | iduronate 2-sulfatase | Ids | AW214631 | 394.0kb | Sense | Chr. X - 70343069 - 70365114 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
