Assay Details
Gene Symbol: Gm12633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033610.1 | 1 | 1552 | 64 | ||
GenBank mRNA | AK135677.1 | - | 1890 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 352 | Zfp352 | 2czf48 | -133.9kb | Anti-sense | Chr. 4 - 90218820 - 90225687 on NCBI Build GRCm38 |
Downstream |
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B) |
Elavl2 |
Hub, mel-N1 |
889.5kb | Sense | Chr. 4 - 91250766 - 91400064 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
