Assay Details
Gene Symbol: E130112N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015604.2 | 2 | 447 | 100 | ||
GenBank mRNA | AK087434.1 | - | 2 | 448 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CD27 antigen | Cd27 |
S152, Tnfrsf7, Tp55 |
-1.6kb | Anti-sense | Chr. 6 - 125232622 - 125237080 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930417O13 gene | 4930417O13Rik | 25.6kb | Sense | Chr. 6 - 125265525 - 125273777 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
