Assay Details
Gene Symbol: Gm12130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040295.1 | 2 - 3 | 305 | 67 | ||
GenBank mRNA | AK145334.1 | - | 2 - 3 | 275 | 67 | |
CJ052615.1 | - | 2 - 3 | 307 | 67 | ||
DV652764.1 | - | 2 - 3 | 249 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 12128 | Gm12128 | OTTMUSG00000005422 | -818.1kb | Anti-sense | Chr. 11 - 37658989 - 37673442 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930553C11 gene | 4930553C11Rik | 812.3kb | Sense | Chr. 11 - 39307703 - 39390225 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361873 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828949 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18202 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17761 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215373 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406485 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36230 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07311222 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07311222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09397339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09508313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725492 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09644775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09508332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725415 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09371587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09281459 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725636 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09397328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09119824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725629 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09553274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725488 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09397333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09586226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09042075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725547 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09042071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725591 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725501 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09044539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09119808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09397330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09397335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09737864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27725435 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1100023524.mm.1 | TC1100002540.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100023524.mm.1 | NR_040295.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100023524.mm.1 | NR_040295.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100023525.mm.1 | TC1100002540.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100023525.mm.1 | NR_040295.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100023525.mm.1 | NR_040295.1 | Refseq |
