Assay Details
Gene Symbol: Gm6602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045362.1 | 1 | 319 | 159 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein arginine N-methyltransferase 6 | Prmt6 |
AW124876, BB233495, Hrmt1l6 |
-1.6mb | Anti-sense | Chr. 3 - 110246104 - 110250999 on NCBI Build GRCm38 |
Downstream | amylase 2b | Amy2b |
Amy-X, Amy2-2, OTTMUSG00000022462, mAmy2-1 |
1.3mb | Anti-sense | Chr. 3 - 113147656 - 113156727 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
