Assay Details
Gene Symbol: Mir376b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029915.1 | 1 | 40 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 654 | Mir654 |
Mirn654, mmu-mir-654 |
-157.0 | Sense | Chr. 12 - 109723218 - 109723301 on NCBI Build GRCm38 |
Downstream | microRNA 376a | Mir376a |
Mirn376a, mir-376a |
242.0 | Sense | Chr. 12 - 109723781 - 109723848 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
