Assay Details
Gene Symbol: 4922502N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045149.1 | 2 - 3 | 171 | 74 | ||
GenBank mRNA | AK015023.1 | - | 2 - 3 | 172 | 74 | |
AK132719.1 | - | 2 - 3 | 172 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | immunoglobulin (CD79A) binding protein 1b | Igbp1b | alpha4-b | -658.5kb | Sense | Chr. 6 - 138657089 - 138658444 on NCBI Build GRCm38 |
Downstream | RERG/RAS-like | Rergl | EG632971 | 172.2kb | Sense | Chr. 6 - 139493182 - 139501909 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
