Assay Details
Gene Symbol: Dnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301737.1 | NP_001288666.1 | 21 - 22 | 2655 | 116 | |
GenBank mRNA | AK220483.1 | - | 21 - 22 | 2597 | 116 | |
L31396.1 | - | 16 - 17 | 1824 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
