Assay Details
Gene Symbol: 1700010I02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040587.1 | 2 - 3 | 210 | 119 | ||
GenBank mRNA | AK005828.1 | - | 2 - 3 | 212 | 119 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | interleukin 7 | Il7 |
A630026I06Rik, Il-7, hlb368 |
-330.1kb | Sense | Chr. 3 - 7572028 - 7613427 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 5330432E05 gene | 5330432E05Rik | ENSMUSG00000058405 | 74.3kb | Sense | Chr. 3 - 8019669 - 8022831 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
