Assay Details
Gene Symbol: Mir6241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105749.1 | 1 | 49 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105245869 | LOC105245869 | -20.3kb | Sense | Chr. 14 - 118235634 - 118238288 on NCBI Build GRCm38 | |
Downstream | predicted gene 9376 | Gm9376 | EG668814 | 8.4kb | Sense | Chr. 14 - 118267158 - 118267779 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
