Assay Details
Gene Symbol: Mir7013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105980.1 | 1 | 32 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | CB946302.1 | - | 1 | Chr. 3 - 95004017 - 95004663 [-] on Build GRCm38 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA B230398E01 gene | B230398E01Rik | 6720476P12Rik | -17.5kb | Anti-sense | Chr. 3 - 94959674 - 94986687 on NCBI Build GRCm38 |
Downstream | zinc finger protein 687 | Zfp687 |
4931408L03Rik, Znf687, mKIAA1441 |
2.5kb | Anti-sense | Chr. 3 - 95006702 - 95015452 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
