Assay Details
Gene Symbol: Mir7656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106116.1 | 1 | 31 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
solute carrier family 9 (sodium/hydrogen exchanger), member 9 |
Slc9a9 |
5730527A11Rik, 9930105B05, AI854429, Nhe9 |
Sense | Chr. 9 - 94669892 - 95230452 on NCBI Build GRCm38 | |
Downstream | carbohydrate sulfotransferase 2 | Chst2 |
AI428561, AW121776, C130041E03Rik, Chts2, GST-2, GlcNAc6ST, Gn6st |
649.0kb | Anti-sense | Chr. 9 - 95400926 - 95407270 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
