Assay Details
Gene Symbol: Gm21859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC082544.1 | - | 3 - 4 | 370 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae) |
Rmi2 |
A630055G03Rik, Gm537, Gm929 |
-68.8kb | Anti-sense | Chr. 16 - 10835059 - 10843235 on NCBI Build GRCm38 |
Downstream | LPS-induced TN factor | Litaf |
3222402J11Rik, C85531, N4WBP3, TBX1 |
45.8kb | Sense | Chr. 16 - 10959273 - 10993121 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
