Assay Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008468.4 | NP_032494.3 | 14 | 5212 | 102 | |
GenBank mRNA | AK154882.1 | - | 14 | 5197 | 102 | |
BC022673.1 | - | 1 | 1321 | 102 | ||
BC048075.1 | - | 1 | 1392 | 102 | ||
BC058586.1 | - | 1 | 1725 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
