Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011680.2 | NP_035810.1 | 10 | 1573 | 113 | |
GenBank mRNA | AK146871.1 | - | 10 | 1520 | 113 | |
AK158906.1 | - | 8 | 1349 | 113 | ||
BC019729.1 | - | 5 | 953 | 113 | ||
BC082995.1 | - | 10 | 1573 | 6440967 | 113 | |
U01662.1 | - | 10 | 1589 | 113 | ||
U01663.1 | - | 7 | 1126 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
