Assay Details
Gene Symbol: Gm21992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290128.1 | NP_001277057.1 | 1 - 2 | 466 | 65 | |
GenBank mRNA | EU287940.1 | - | 1 - 2 | 340 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
