Assay Details
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 12 - 13 | 1824 | 102 | |
GenBank mRNA | AK030158.1 | - | 4 - 5 | 523 | 102 | |
L11625.1 | - | 3 - 4 | 362 | 102 | ||
U21301.1 | - | 12 - 13 | 1824 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
