Assay Details
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 17 - 18 | 2386 | 107 | |
GenBank mRNA | AK029009.1 | - | 6 - 7 | 588 | 107 | |
AK030158.1 | - | 9 - 10 | 1085 | 107 | ||
L11625.1 | - | 8 - 9 | 924 | 107 | ||
U21301.1 | - | 17 - 18 | 2386 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
