Assay Details
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029250.2 | NP_083526.2 | 4 - 5 | 949 | 103 | |
GenBank mRNA | AK016135.1 | - | 4 - 5 | 931 | 103 | |
AK031560.1 | - | 926 | 103 | |||
AK033268.1 | - | 4 - 5 | 926 | 103 | ||
AK044502.1 | - | 4 - 5 | 950 | 103 | ||
AK049827.1 | - | 4 - 5 | 663 | 103 | ||
AK051039.1 | - | 959 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
