Assay Details
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042542.2 | NP_001036007.1 | 1 - 2 | 148 | 79 | |
GenBank mRNA | CJ047282.1 | - | 1 - 2 | 146 | 79 | |
U07423.1 | - | 1 - 2 | 117 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001042542.2, U07423.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
