Assay Details
Gene Symbol: Als2cr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175200.4 | NP_780409.2 | 10 - 11 | 1242 | 172 | |
GenBank mRNA | AK076617.1 | - | 10 - 11 | 1244 | 172 | |
BC049729.1 | - | 6 - 7 | 459 | 172 | ||
BC118929.2 | - | 10 - 11 | 1208 | 40090362 | 172 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
