Assay Details
Gene Symbol: Rassf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175279.3 | NP_780488.2 | 1 | 1122 | 66 | |
GenBank mRNA | AK019489.1 | - | 1 | 569 | 66 | |
AK031660.1 | - | 1 | 1123 | 66 | ||
AK046433.1 | - | 1 | 1118 | 66 | ||
BC028854.1 | - | 1 | 698 | 66 | ||
BC125291.1 | - | 1 | 938 | 40129806 | 66 | |
BC125293.1 | - | 1 | 938 | 40129808 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
