Assay Details
Gene Symbol: Glo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113560.1 | NP_001107032.1 | 4 - 5 | 486 | 79 | |
NM_025374.3 | NP_079650.3 | 4 - 5 | 486 | 79 | ||
GenBank mRNA | AK002386.1 | - | 4 - 5 | 414 | 79 | |
AK003567.1 | - | 4 - 5 | 434 | 79 | ||
AK005055.1 | - | 4 - 5 | 469 | 79 | ||
AK005215.1 | - | 4 - 5 | 435 | 79 | ||
AK031832.1 | - | 4 - 5 | 428 | 79 | ||
AK049703.1 | - | 4 - 5 | 443 | 79 | ||
BC024663.1 | - | 4 - 5 | 445 | 3591599 | 79 | |
BC081432.1 | - | 4 - 5 | 440 | 6819667 | 79 | |
BG077218.2 | - | 4 - 5 | 457 | 79 | ||
DQ487161.1 | - | 4 - 5 | 405 | 79 | ||
DQ487162.1 | - | 4 - 5 | 405 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
