Assay Details
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009319.2 | NP_001009319.1 | 5 - 6 | 608 | 75 | |
GenBank mRNA | S67450.1 | - | 5 - 6 | 608 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
