Assay Details
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970078.1 | XP_027825879.1 | 2 - 3 | 1848 | 71 | |
XM_027970079.1 | XP_027825880.1 | 3 - 4 | 1705 | 71 | ||
XM_027970080.1 | XP_027825881.1 | 3 - 4 | 1312 | 71 | ||
XM_027970082.1 | XP_027825883.1 | 3 - 4 | 347 | 71 | ||
XM_027970083.1 | XP_027825884.1 | 3 - 4 | 322 | 71 | ||
XM_027970084.1 | XP_027825885.1 | 2 - 3 | 258 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
