Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017363.3 | XP_004017412.1 | 2 - 3 | 294 | 72 | |
XM_027956502.1 | XP_027812303.1 | 3 - 4 | 296 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
