Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017363.3 | XP_004017412.1 | 3 - 4 | 526 | 89 | |
XM_027956502.1 | XP_027812303.1 | 4 - 5 | 528 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
