Assay Details
Gene Symbol: BCL2L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002344.4 | XP_004002393.1 | 4 - 5 | 680 | 76 | |
XM_027973353.1 | XP_027829154.1 | 3 - 4 | 435 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
