Assay Details
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127672.1 | NP_001121144.1 | 3 - 4 | 519 | 100 | |
GenBank mRNA | EU650274.1 | - | 3 - 4 | 519 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
