Assay Details
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127672.1 | NP_001121144.1 | 4 - 5 | 586 | 94 | |
GenBank mRNA | EU650274.1 | - | 4 - 5 | 586 | 94 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AKIRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127672.1 | NP_001121144.1 | 4 - 5 | 586 | 94 | |
GenBank mRNA | EU650274.1 | - | 4 - 5 | 586 | 94 |