Assay Details
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246263.2 | NP_001233192.2 | 2 - 3 | 744 | 98 | |
GenBank mRNA | HQ678172.1 | - | 2 - 3 | 471 | 98 | |
JQ941956.1 | - | 2 - 3 | 744 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
