Assay Details
Gene Symbol: LOC101110521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973621.1 | XP_027829422.1 | 14 - 15 | 2193 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101110521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973621.1 | XP_027829422.1 | 14 - 15 | 2193 | 115 |
Set Membership: |
![]() ![]() ![]() ![]() |