Assay Details
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092271.2 | XP_014947757.2 | 3 - 4 | 259 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092271.2 | XP_014947757.2 | 3 - 4 | 259 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |