Assay Details
Gene Symbol: TTC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017022.3 | XP_004017071.1 | 3 - 4 | 363 | 69 | |
XM_012097004.2 | XP_011952394.1 | 3 - 4 | 511 | 69 | ||
XM_012097005.2 | XP_011952395.1 | 3 - 4 | 452 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
