Assay Details
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964976.1 | XP_027820777.1 | 4 - 5 | 1068 | 68 | |
XM_027964977.1 | XP_027820778.1 | 2 - 3 | 847 | 68 | ||
XM_027964978.1 | XP_027820779.1 | 4 - 5 | 1068 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
